Merck
  • Pericytes impair capillary blood flow and motor function after chronic spinal cord injury.
Pericytes impair capillary blood flow and motor function after chronic spinal cord injury.
Nature medicine (2017-05-02)
Yaqing Li, Ana M Lucas-Osma, Sophie Black, Mischa V Bandet, Marilee J Stephens, Romana Vavrek, Leo Sanelli, Keith K Fenrich, Antonio F Di Narzo, Stella Dracheva, Ian R Winship, Karim Fouad, David J Bennett
ABSTRACT
Blood vessels in the central nervous system (CNS) are controlled by neuronal activity. For example, widespread vessel constriction (vessel tone) is induced by brainstem neurons that release the monoamines serotonin and noradrenaline, and local vessel dilation is induced by glutamatergic neuron activity. Here we examined how vessel tone adapts to the loss of neuron-derived monoamines after spinal cord injury (SCI) in rats. We find that, months after the imposition of SCI, the spinal cord below the site of injury is in a chronic state of hypoxia owing to paradoxical excess activity of monoamine receptors (5-HT1) on pericytes, despite the absence of monoamines. This monoamine-receptor activity causes pericytes to locally constrict capillaries, which reduces blood flow to ischemic levels. Receptor activation in the absence of monoamines results from the production of trace amines (such as tryptamine) by pericytes that ectopically express the enzyme aromatic L-amino acid decarboxylase (AADC), which synthesizes trace amines directly from dietary amino acids (such as tryptophan). Inhibition of monoamine receptors or of AADC, or even an increase in inhaled oxygen, produces substantial relief from hypoxia and improves motoneuron and locomotor function after SCI.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Serotonin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Donkey Anti-Sheep IgG Antibody, biotin-SP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Anti-von Willebrand Factor Antibody, Chemicon®, from rabbit