- Bufalin inhibits cell proliferation and migration of hepatocellular carcinoma cells via APOBEC3F induced intestinal immune network for IgA production signaling pathway.
Bufalin inhibits cell proliferation and migration of hepatocellular carcinoma cells via APOBEC3F induced intestinal immune network for IgA production signaling pathway.
This study aimed to evaluate functions of APOBEC3F gene in biological process of hepatocellular carcinoma (HCC) and anti-tumor mechanisms of bufalin. Effect of APOBEC3F and bufalin on cell proliferation and migration abilities were evaluated by CCK-8, wounding healing tests and transwell assays in SK-Hep1 and Bel-7404āÆcells. Bioinformatic analysis were also used to compare APOBEC3F expression levels, detect coexpressed genes and enrichment of pathways. APOBEC3F was overexpressed in tumor tissues compared to adjacent tissues in HCC patients. And, APOBEC3F promotes cell proliferation and migration in SK-Hep1 and Bel-7404āÆcells. Bufalin inhibits cell proliferation and migration and reduces APOBEC3F expression. GO and KEGG enrichment of APOBEC3F-coexpressed genes revealed that APOBEC3F might active intestinal immune network for IgA production signaling pathway, leading to malignant biological behaviors of HCC cells. Additionally, siAPOBEC3F could decrease pIgR, CCR9, CCR10 and CXCR4 protein levels. And, bufalin inhibits the pIgR, CCR9, CCR10 and CXCR4 protein expressions. Bufalin inhibits cell proliferation and migration of HCC cells via APOBEC3F induced intestinal immune network for IgA production signaling pathway.