
Yttrium(III) nitrate hexahydrate

Synonym(s):
Yttrium(III) nitrate hexahydrate
Linear Formula:
Y(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
383.01
Products
Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Reagent Type
Core
Showing 1-3 of 3