Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Water solution
Linear Formula:
H
2
O
Products
Product Category
solvents (6)
Brand
Sigma-Aldrich (5)
Supelco (1)
Physical Form
Quality Segment
Application
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
576905
Water solution,
contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Expand
View Pricing
576913
Water solution,
suitable for HPLC, contains 0.1 % (v/v) formic acid
Expand
View Pricing
679178
Water solution,
contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid
Expand
View Pricing
667404
Water solution,
contains 10 mM ammonium acetate, suitable for HPLC
Expand
View Pricing
900687
Water solution,
contains 0.1 % (v/v) formic acid, for UHPLC, suitable for mass spectrometry (MS)
Expand
View Pricing
633321
Water solution,
contains 0.1 % (v/v) formic acid, 20 % (v/v) acetonitrile
Expand
View Pricing