Vicenin II
Synonym(s):
Vicenin 2, 6,8-Di-C-β-D-glucopyranosylapigenin, Apigenin 6,8-C-di-β-D-glucopyranoside, Apigenin 6,8-diglucoside
Empirical Formula (Hill Notation):
C27H30O15
CAS Number:
Molecular Weight:
594.52
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1