
Trimethyl(methylcyclopentadienyl)platinum(IV)

Linear Formula:
C5H4CH3Pt(CH3)3
CAS Number:
Molecular Weight:
319.30
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing