![Tetrachloroethylene Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/188/998/32a663fe-5820-47cb-86d7-4239f6be8778/640/32a663fe-5820-47cb-86d7-4239f6be8778.png)
Tetrachloroethylene
![Tetrachloroethylene Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/188/998/32a663fe-5820-47cb-86d7-4239f6be8778/640/32a663fe-5820-47cb-86d7-4239f6be8778.png)
Synonym(s):
Tetrachloroethylene, PCE, Perchloroethylene
Linear Formula:
CCl2=CCl2
CAS Number:
Molecular Weight:
165.83
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing