
Swertiamarine

Synonym(s):
Swertiamarin, (4aR,5R,6S)-5-Ethenyl-6-(β-D-glucopyranosyloxy)-4,4a,5,6-tetrahydro-4a-hydroxy1H,3H-pyrano[3,4-c]pyran-1-one, 4,4a,5,6-Tetrahydro-4aα-hydroxy-1-oxo-5β-vinyl-1H,3H-pyrano[3,4-c]pyran-6-yl β-D-glucopyranoside, Swertiamarine, Swertiamaroside
Empirical Formula (Hill Notation):
C16H22O10
CAS Number:
Molecular Weight:
374.34
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing