
Sodium persulfate

Synonym(s):
Sodium peroxodisulfate
Linear Formula:
Na2S2O8
CAS Number:
Molecular Weight:
238.10
Products
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Special Grade