![(−)-Sinigrin hydrate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/374/668/38ad48ef-f02d-41f9-a90c-763ae8f9ed46/640/38ad48ef-f02d-41f9-a90c-763ae8f9ed46.png)
(−)-Sinigrin hydrate
![(−)-Sinigrin hydrate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/374/668/38ad48ef-f02d-41f9-a90c-763ae8f9ed46/640/38ad48ef-f02d-41f9-a90c-763ae8f9ed46.png)
Empirical Formula (Hill Notation):
C10H16KNO9S2 · xH2O
CAS Number:
Molecular Weight:
397.46 (anhydrous basis)
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing