
Salbutamol impurity B

Synonym(s):
(1RS)-2-[(1,1-Dimethylethyl)amino]-1-(4-hydroxyphenyl)ethanol, 2-tert-Butylamino-1-(4-hydroxyphenyl)ethanol
Empirical Formula (Hill Notation):
C12H19NO2
CAS Number:
Molecular Weight:
209.28
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade