
Pseudoisoeugenyl 2-methylbutyrate for peak identification

Synonym(s):
Anise oil, Anise seed oil
CAS Number:
Products
Boiling Point (°C)
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing