Skip to Content
Merck
Potassium trifluoromethanesulfonate 98%

Potassium trifluoromethanesulfonate

Potassium trifluoromethanesulfonate 98%
Synonym(s):
Potassium triflate, Trifluoromethanesulfonic acid potassium salt
Linear Formula:
CF3SO3K
CAS Number:
Molecular Weight:
188.17

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing