
Potassium phosphate dibasic solution

Synonym(s):
Dipotassium phosphate
Empirical Formula (Hill Notation):
HK2O4P
CAS Number:
Molecular Weight:
174.18
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Quality Segment
Application
Sterilization Method
Showing 1-1 of 1
Compare
Product Number
Description
Pricing