
Phyllanthin

Synonym(s):
(2S,3S)-(+)-1,4-Dimethoxy-2,3-diveratrylbutane
Empirical Formula (Hill Notation):
C24H34O6
CAS Number:
Molecular Weight:
418.52
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing