
Oxycodone Related Compound C

Synonym(s):
Codeinone, 6-Oxocodeine, 7,8-Didehydro-4,5α-epoxy-3-methoxy-17-methylmorphinan-6-one
Empirical Formula (Hill Notation):
C18H19NO3
CAS Number:
Molecular Weight:
297.35
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2