Skip to Content
Merck
O-(Trimethylsilyl)hydroxylamine technical grade, 90%

O-(Trimethylsilyl)hydroxylamine

O-(Trimethylsilyl)hydroxylamine technical grade, 90%
Synonym(s):
Aminoxytrimethylsilane
Linear Formula:
(CH3)3SiONH2
CAS Number:
Molecular Weight:
105.21

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing