Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
-(Phosphonomethyl)glycine
Synonym(s):
Glyphosate
Linear Formula:
(HO)
2
P(O)CH
2
NHCH
2
CO
2
H
CAS Number:
1071-83-6
Molecular Weight:
169.07
EC Number:
213-997-4
Products
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
337757
N
-(Phosphonomethyl)glycine,
96%, for peptide synthesis
Expand
View Pricing
P9556
N
-(Phosphonomethyl)glycine,
BioReagent, suitable for plant cell culture
Expand
View Pricing