
Niobium(V) chloride

Synonym(s):
Niobium pentachloride, Niobium(V) chloride
Linear Formula:
NbCl5
CAS Number:
Molecular Weight:
270.17
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Technique
Showing 1-4 of 4