Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel/Boron/Silicon
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
GF63130070
Nickel/Boron/Silicon,
foil, thickness 0.025 mm, Ni78%/B 14%/Si 8%, length 1 m, coil width 25 mm
Expand
View Pricing
GF91259141
Nickel/Boron/Silicon,
foil, thickness 0.025 mm, Ni78%/B 14%/Si 8%, length 2 m, coil width 25 mm
Expand
View Pricing
GF23015359
Nickel/Boron/Silicon,
foil, Ni78%/B 14%/Si 8, thickness 0.04 mm, length 2 m, coil width 25 mm
Expand
View Pricing
GF14895625
Nickel/Boron/Silicon,
foil, Ni78%/B 14%/Si 8, thickness 0.04 mm, length 10 m, coil width 25 mm
Expand
View Pricing
GF24168416
Nickel/Boron/Silicon,
foil, Ni78%/B 14%/Si 8, thickness 0.025 mm, length 10 m, coil width 25 mm
Expand
View Pricing
GF68505048
Nickel/Boron/Silicon,
foil, Ni78%/B 14%/Si 8%, thickness 0.025 mm, length 0.5 m, coil width 25 mm
Expand
View Pricing
GF07393349
Nickel/Boron/Silicon,
foil, Ni78%/B 14%/Si 8, thickness 0.025 mm, length 5 m, coil width 25 mm
Expand
View Pricing
GF38868676
Nickel/Boron/Silicon,
foil, Ni78%/B 14%/Si 8, thickness 0.04 mm, length 0.5 m, coil width 25 mm
Expand
View Pricing
GF58584123
Nickel/Boron/Silicon,
foil, Ni78%/B 14%/Si 8, thickness 0.04 mm, length 1 m, coil width 25 mm
Expand
View Pricing