
Naproxen Impurity C

Synonym(s):
(2S)-2-(5-Bromo-6-methoxynaphthalen-2-yl)propanoic acid, (αS)-5-Bromo-6-methoxy-α-methyl-2-naphthaleneacetic acid, (2S)-2-(5-Bromo-6-methoxy-2-naphthyl)propionic acid, (S)-5-Bromonaproxen
Empirical Formula (Hill Notation):
C14H13BrO3
CAS Number:
Molecular Weight:
309.16
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing