Skip to Content
Merck
vibrant-m

N-(2-Hydroxyethyl)isonicotinamide nitric ester

vibrant-m
Synonym(s):
2-[(Pyridin-4-ylcarbonyl)amino]ethyl nitrate, p-Nicorandil
Empirical Formula (Hill Notation):
C8H9N3O4
CAS Number:
Molecular Weight:
211.17

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1