Skip to Content
Merck
Myclobutanil PESTANAL®, analytical standard

Myclobutanil

Myclobutanil PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C15H17ClN4
CAS Number:
Molecular Weight:
288.78

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique