
Monocrotaline

Synonym(s):
Crotaline, Monocrotaline
Empirical Formula (Hill Notation):
C16H23NO6
CAS Number:
Molecular Weight:
325.36
Products
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing