
Methyl ricinoleate

Synonym(s):
(R)-12-Hydroxy-cis-9-octadecenoic acid methyl ester, Methyl 12-hydroxyoleate, Ricinoleic acid methyl ester, Methyl ricinoleate
Empirical Formula (Hill Notation):
C19H36O3
CAS Number:
Molecular Weight:
312.49
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing