
Methyl caprylate

Synonym(s):
Methyl octanoate, Caprylic acid methyl ester, Methyl caprylate, Octanoic acid methyl ester
Linear Formula:
CH3(CH2)6COOCH3
CAS Number:
Molecular Weight:
158.24
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing