Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methanol-OD
Synonym(s):
mono-Deuteromethanol, Methan(ol-d), Methyl alcohol-OD
Linear Formula:
CH
3
OD
CAS Number:
1455-13-6
Molecular Weight:
33.05
Products
Product Category
alcohols (2)
solvents (2)
stable isotopes (2)
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Isotope
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
151939
Methanol-OD,
99.5 atom % D
Expand
View Pricing
550574
Methanol-OD,
99 atom % D
Expand
View Pricing
900843
Methanol-OD,
reagent grade, ≥99 atom % D
Expand
View Pricing
793345
Methanol-OD,
99 atom % D, contains 2 mg/mL 1-butanol-d
10
(99 atom % D), 2 mg/mL 2-propanol-1,1,1,3,3,3-d
6
(99 atom % D)
Expand
View Pricing