Skip to Content
Merck
Memantine Hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material

Memantine hydrochloride

Memantine Hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
3,5-Dimethyl-1-adamantanamine hydrochloride, 3,5-Dimethylamantadine hydrochloride, 3,5-Dimethyltricyclo[3.3.1.13,7]decan-1-amine hydrochloride, Memantine hydrochloride
Empirical Formula (Hill Notation):
C12H21N · HCl
CAS Number:
Molecular Weight:
215.76
EC Number:

Products

Product Category
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique