L-Methionine-1-13C
Linear Formula:
CH3SCH2CH2CH(NH2)13CO2H
CAS Number:
Molecular Weight:
150.20
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing