
Lithium tert-butoxide solution

Linear Formula:
(CH3)3COLi
CAS Number:
Molecular Weight:
80.05
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing