Skip to Content
Merck
Lithium iron(II) phosphate powder, <5 μm particle size (BET), >97% (XRF)

Lithium iron(II) phosphate

Lithium iron(II) phosphate powder, <5 μm particle size (BET), >97% (XRF)
Synonym(s):
Ferrous lithium phosphate, Iron lithium phosphate, LFP, Triphylite
Linear Formula:
LiFePO4
CAS Number:
Molecular Weight:
157.76

Products

Product Category
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Application
Particle Size (µm)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing