
L-Ethionine

Synonym(s):
L-2-Amino-4-(ethylthio)butyric acid
Linear Formula:
C2H5SCH2CH2CH(NH2)CO2H
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing