Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Isopentyl acetate
Synonym(s):
Acetic acid 3-methylbutyl ester, Isoamyl acetate
Linear Formula:
CH
3
COOCH
2
CH
2
CH(CH
3
)
2
CAS Number:
123-92-2
Molecular Weight:
130.18
EC Number:
204-662-3
Products
Product Category
solvents (2)
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Special Grade
Anhydrous (1)
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
112674
Isopentyl acetate,
mixture of isomers, reagent grade, 98%
Expand
View Pricing
306967
Isopentyl acetate,
anhydrous, ≥99%
Expand
View Pricing
79857
Isopentyl acetate,
analytical standard
Expand
View Pricing
15-1560
Isopentyl acetate,
SAJ special grade, ≥98.0%
Expand
View Pricing