
Iron oxide(II,III), magnetic nanoparticles solution

Synonym(s):
Magnetic iron oxide nanocrystals, Magnetite, Superparamagnetic iron oxide nanoparticles
Empirical Formula (Hill Notation):
Fe3O4
CAS Number:
Molecular Weight:
231.53
Products
Brand
Color
Formula Weight
Physical Form
Functional Group
Showing 1-13 of 13
Compare
Product Number
Description
Pricing