
Ibuprofen Related Compound J

Synonym(s):
(2RS)-2-(4-Isobutyrylphenyl)propionic acid, α-Methyl-4-(2-methyl-1-oxopropyl)benzeneacetic acid, (2RS)-2-[4-(2-Methylpropanoyl)phenyl]propanoic acid, 1-Oxoibuprofen
Empirical Formula (Hill Notation):
C13H16O3
CAS Number:
Molecular Weight:
220.26
Products
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2