Skip to Content
Merck
Haloxyfop-methyl PESTANAL®, analytical standard

Haloxyfop-methyl

Haloxyfop-methyl PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C16H13ClF3NO4
CAS Number:
Molecular Weight:
375.73

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique