Formestane
Synonym(s):
4-Hydroxyandrost-4-ene-3,17-dione, CGP-32349
Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
Products
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1