
Fomesafen

Synonym(s):
5-[2-Chloro-4-(trifluoromethyl)phenoxy]-N-(methylsulfonyl)-2-nitrobenzamide
Empirical Formula (Hill Notation):
C15H10ClF3N2O6S
CAS Number:
Molecular Weight:
438.76
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2