
Flurbiprofen

Synonym(s):
(±)-2-Fluoro-α-methyl-4-biphenylacetic acid, L-790,330, Flurbiprofen
Linear Formula:
C6H5C6H3(F)CH(CH3)CO2H
CAS Number:
Molecular Weight:
244.26
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing