Ethyl 3-(trimethylsilyl)propynoate
Synonym(s):
Ethyl 3-(trimethylsilyl)propiolate
Linear Formula:
(CH3)3SiC≡CCO2C2H5
CAS Number:
Molecular Weight:
170.28
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing