Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Safe-Lock Microcentrifuge Tubes, Biopur
®
Products
Product Category
tubes (3)
Color
Manufacturer
Volume (ml)
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
SLMTBP15-EP
Eppendorf
®
Safe-Lock Microcentrifuge Tubes, Biopur
®
,
volume 1.5 mL, colorless, pkg of 100 ea (individually wrapped)
Expand
View Pricing
SLMTBP2-EP
Eppendorf
®
Safe-Lock Microcentrifuge Tubes, Biopur
®
,
volume 2.0 mL, colorless, pkg of 100 ea (individually wrapped)
Expand
View Pricing
SLMTBP5-EP
Eppendorf
®
Safe-Lock Microcentrifuge Tubes, Biopur
®
,
volume 0.5 mL, colorless, pkg of 50 ea (individually wrapped)
Expand
View Pricing