Skip to Content
Merck
D-Lactose monohydrate SAJ special grade

D-Lactose monohydrate

D-Lactose monohydrate SAJ special grade
Synonym(s):
β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, Milk sugar
Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
EC Number:

Products

Product Category
Brand
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Optical Isomer
Special Grade