Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(±)-1,3-Butanediol
Synonym(s):
1,3-Butylene glycol
Linear Formula:
CH
3
CH(OH)CH
2
CH
2
OH
CAS Number:
107-88-0
Molecular Weight:
90.12
EC Number:
203-529-7
Products
Product Category
building blocks (1)
solvents (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Anhydrous (1)
analytical standard (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
309443
(±)-1,3-Butanediol,
anhydrous, ≥99%
Expand
View Pricing
B84785
(±)-1,3-Butanediol,
ReagentPlus
®
, 99.5%
Expand
View Pricing
09068
(±)-1,3-Butanediol,
analytical standard
Expand
View Pricing
03-4450
(±)-1,3-Butanediol,
SAJ first grade, ≥98.0%
Expand
View Pricing