
Di(trimethylolpropane)

Synonym(s):
2,2′-Oxybis(methylene)bis(2-ethyl-1,3-propanediol)
Linear Formula:
O[CH2C(C2H5)(CH2OH)2]2
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing