Skip to Content
Merck
Di-tert-butyl dicarbonate ≥98.0% (GC)

Di-tert-butyl dicarbonate

Di-tert-butyl dicarbonate ≥98.0% (GC)
Synonym(s):
Boc anhydride, Di-tert-butyl pyrocarbonate, Di-tert-butyl dicarbonate
Linear Formula:
[(CH3)3COCO]2O
CAS Number:
Molecular Weight:
218.25
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reaction Type