
Di(propylene glycol) methyl ether, mixture of isomers

Synonym(s):
Dipropylene glycol monomethyl ether
Linear Formula:
CH3OC3H6OC3H6OH
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Polymer Composition
Polymer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing