
Diosgenin

Synonym(s):
(25R)-5-Spirosten-3β-ol, 3β-Hydroxy-5-spirostene, Nitogenin
Empirical Formula (Hill Notation):
C27H42O3
CAS Number:
Molecular Weight:
414.62
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing