![Dihydrorhodamine 123 ≥95%](/deepweb/assets/sigmaaldrich/product/structures/195/084/4f7c2785-b3ae-4217-91ef-9d0886615cbd/640/4f7c2785-b3ae-4217-91ef-9d0886615cbd.png)
Dihydrorhodamine 123
![Dihydrorhodamine 123 ≥95%](/deepweb/assets/sigmaaldrich/product/structures/195/084/4f7c2785-b3ae-4217-91ef-9d0886615cbd/640/4f7c2785-b3ae-4217-91ef-9d0886615cbd.png)
Synonym(s):
Dihydrorhodamine 123
Empirical Formula (Hill Notation):
C21H18N2O3
CAS Number:
Molecular Weight:
346.38
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Technique
Showing 1-2 of 2