Skip to Content
Merck
Dihydrocodeine Bitartrate certified reference material, pharmaceutical secondary standard

Dihydrocodeine bitartrate

Dihydrocodeine Bitartrate certified reference material, pharmaceutical secondary standard
Synonym(s):
Dihydrocodeine hydrogen tartrate
Empirical Formula (Hill Notation):
C18H23NO3 · C4H6O6
CAS Number:
Molecular Weight:
451.47

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade