
Desosaminylazithromycin

Synonym(s):
Desosaminylazithromycin
Empirical Formula (Hill Notation):
C30H58N2O9
CAS Number:
Molecular Weight:
590.79
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2